Learn More
Bachem Secretin (human)
SDP
Supplier:  Bachem 4031250.0001
Encompass_Preferred
Secretin (human) , H-His-Ser-Asp-Gly-Thr-Phe-Thr-Ser-Glu-Leu-Ser-Arg-Leu-Arg-Glu-Gly-Ala-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Gly-Leu-Val-NH2, 108153-74-8, C130H220N44O40, Mr 3039.45, 1mg. Gastrointestinal hormone which stimulates pancreatic secretion.
Catalog No. 50-259-686