Learn More
Cusabio Technology LLC secretory immunoglobulin A
Catalog No. 5000049313
Encompass_Preferred
This kit detects sIgA in rat serum plasma and tissue homogenates offering a quantitative measurement within a range of 0 78 ng/mL to 50 ng/mL and a sensitivity of 0 195 ng/mL Based on a sandwich ELISA methodology it enables precise analysis with an incubation period of 1-5 hours and a sample volume requirement of 50-100 L The assay utilizes absorbance readings at 450 nm to determine sIgA concentrations ensuring reliable results for evaluating secretory immunoglobulin A levels in diverse biological samples Its standardized protocol supports efficient workflow compatibility for research applications requiring accurate quantification
Catalog No. 50-000-49313 Supplier Cusabio Technology LLC Supplier No. CSBE08412R
This kit detects sIgA in rat serum plasma and tissue homogenates offering a quantitative measurement within a range of 0 78 ng/mL to 50 ng/mL and a sensitivity of 0 195 ng/mL Based on a sandwich ELISA methodology it enables precise analysis with an incubation period of 1-5 hours and a sample volume requirement of 50-100 L The assay utilizes absorbance readings at 450 nm to determine sIgA concentrations ensuring reliable results for evaluating secretory immunoglobulin A levels in diverse biological samples Its standardized protocol supports efficient workflow compatibility for research applications requiring accurate quantification