Learn More
Electron Microscopy Sciences Section Lifter Model A
SDP
Catalog No. 5028537
Encompass_Preferred
Made from polished stainless steel. Blade measures 17/8"x1.5" (4.8x3.8cm), overall length is 6" (15.2cm)
Catalog No. 50-285-37 Supplier Electron Microscopy Sciences Supplier No. 6307610
Made from polished stainless steel. Blade measures 17/8"x1.5" (4.8x3.8cm), overall length is 6" (15.2cm)