Learn More
Sino Biological SECTM1 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 12269MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SECTM1 (rh SECTM1; Catalog#12269-H08H; Q8WVN6-1; Met1-Gly145). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8680