Learn More
Sino Biological SECTM1 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 12269R106100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SECTM1 (rh SECTM1; Catalog#12269-H08H; Q8WVN6-1; Met1-Gly145).
Catalog No. 50-224-7905