Learn More
Medchemexpress LLC HY-P9927 1mg , Secukinumab CAS:1229022-83-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP99271MG
Encompass_Preferred
HY-P9927 1mg Secukinumab CAS: 1229022-83-6 Secukinumab (AIN457) is a high affinity, human monoclonal antibody targeted against interleukin (IL)-17A. Secukinumab is the first-in-class anti-IL-17 agent used for the research of plaque psoriasis, ankylosing spondylitis and psoriatic arthritis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1590