Learn More
Martor Secunorm 525 W/ Rounded Tip Blade
Catalog No. NC1955862
Encompass_Preferred
The SECUNORM 525 is a straightforward safety knife in every way. The ergonomic aluminum handle sits firmly and comfortably in the hand. Its robust design and large cutting depth of up to 18 mm means the SECUNORM 525 easily manages a variety of cutting materials, for example 3-ply cardboard. An unobstructed view of the blade also allows you to cut particularly precisely. In every task, the proven automatic blade retraction protects you from cutting injuries.
Catalog No. NC1955862 Supplier Martor Supplier No. 52500110.02
The SECUNORM 525 is a straightforward safety knife in every way. The ergonomic aluminum handle sits firmly and comfortably in the hand. Its robust design and large cutting depth of up to 18 mm means the SECUNORM 525 easily manages a variety of cutting materials, for example 3-ply cardboard. An unobstructed view of the blade also allows you to cut particularly precisely. In every task, the proven automatic blade retraction protects you from cutting injuries.