Learn More
Martor Secuplus Rippex 210, Replaces #9.08 ORIGINAL Ripper (small)
Catalog No. NC3804316
Encompass_Preferred
The SECUPLUS RIPPEX 210 is ideal for processing roll material. Thanks to a stable edge on the top, it is able to split individual layers of paper from the roll. Since there is no blade, cutting injuries are impossible. And material shrinkage is also kept within limits. Because the ergonomic SECUPLUS RIPPEX 210 can not only be inserted precisely into the selected layer. It also glides over the surface without leaving any marks.-Bladeless cutter for maximum safety-Ergonomic Handle-Beveled Edge-Smooth Contact Surface-Replaces #9.08 ORIGINAL Ripper (small)
Catalog No. NC3804316 Supplier Martor Supplier No. 210001.13
The SECUPLUS RIPPEX 210 is ideal for processing roll material. Thanks to a stable edge on the top, it is able to split individual layers of paper from the roll. Since there is no blade, cutting injuries are impossible. And material shrinkage is also kept within limits. Because the ergonomic SECUPLUS RIPPEX 210 can not only be inserted precisely into the selected layer. It also glides over the surface without leaving any marks.-Bladeless cutter for maximum safety-Ergonomic Handle-Beveled Edge-Smooth Contact Surface-Replaces #9.08 ORIGINAL Ripper (small)