Learn More
Martor Secupro 625
Catalog No. NC1080324
Encompass_Preferred
The Secupro 625 is the ultimate safety knife. It's easy and safe to use yet durable and features an inch of blade extension. It's the most versatile safety knife on the market because it works great for light materials up to heavy duty cuts and everything in between.- Fully automatic retractable blade for safety- Ergonomically friendly trigger mechanism to engage the blade- Durable body made of aluminum with soft touch rubber for grip- Safer alternative to fixed blade utility knives for general cutting tasks
Catalog No. NC1080324 Supplier Martor Supplier No. 625001.02
The Secupro 625 is the ultimate safety knife. It's easy and safe to use yet durable and features an inch of blade extension. It's the most versatile safety knife on the market because it works great for light materials up to heavy duty cuts and everything in between.- Fully automatic retractable blade for safety- Ergonomically friendly trigger mechanism to engage the blade- Durable body made of aluminum with soft touch rubber for grip- Safer alternative to fixed blade utility knives for general cutting tasks