Learn More
3M™ Secure Click™ Full Facepiece Reusable Respirator
Catalog No. 17100822 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-100-822 Large
17-100-821 Small
17-100-820 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-100-822 Supplier 3M™ Supplier No. 7100261328
Add to Cart
Add to Cart
The Secure Click™ Full Facepiece Reusable Respirator helps protect against particulates and/or a wide variety of gases and vapors when used with approved Secure Click™ filters and cartridges.
  • Facepiece comes with ideal features that make it 3M™'s next generation in comfort, breathability and simplicity
TRUSTED_SUSTAINABILITY
Specifications
Certifications (NIOSH) NIOSH Approved
Color Black
Product Type Respirator
Size Large
With Exhalation Valve No
Includes 4 Respirators
Latex Free Yes
Neck Strap No
No. of Ports Dual