Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Secure Click™ Half Facepiece Parts and Accessories

Catalog No. p-7229353 Shop All 3M Products
Change view
Click to view available options
Product Type:
Inhale and Exhale Valve
Speaking Diaphragm Assembly
Includes:
2 Replacement Valves/Facepiece
5 Replacement Speaking Diaphragms/ Case
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Includes
19-072-227 Speaking Diaphragm Assembly 5 Replacement Speaking Diaphragms/ Case
19-072-228 Inhale and Exhale Valve 2 Replacement Valves/Facepiece
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072-227 Supplier 3M™ Supplier No. 7100181934
Only null left
Add to Cart
Add to Cart

For use with Secure Click™ half facepiece respirators.

  • Replacement part for 3M™ Secure Click™ Half Facepiece Reusable Respirator HF-800 Series
  • For workplace and occupational uses only
TRUSTED_SUSTAINABILITY

Specifications

Description Accessory
Color Black, Yellow
Material Nylon
Product Type Speaking Diaphragm Assembly
For Use With (Equipment) Secure Click™ HF-800 Series Respirators
Includes 5 Replacement Speaking Diaphragms/ Case
Model HF-800-05
Quantity 5/Cs.
Dimensions (L x W x H) 12.4 x 42.8 x 42.8 mm
Weight (Metric) 5.44 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.