Learn More
3M™ Secure Click™ Quantitative Fit Test Adapter Kit
Catalog No. 19072226 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
3M™ Secure Click™ HF-800 Series Half Facepiece Reusable Respirators
Catalog No. For Use With (Equipment)
19-072-226 3M™ Secure Click™ HF-800 Series Half Facepiece Reusable Respirators
1 options
Catalog No. 19-072-226 Supplier 3M™ Supplier No. 7100181558
Add to Cart
Add to Cart
Quantitative fit test adapter kit enables fit testing on HF-800 Series half-mask respirators.
  • Use to conduct quantitative fit testing on 3M™ Secure Click™ Half Facepiece Respirators HF-800 Series
  • Includes head harness assembly and fit test adapter with port
  • HF-800 Series facepieces and Secure Click™ cartridges and filters are not included
  • For workplace and occupational use only
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fit Test Adapter
For Use With (Application) Quantitative Fit Testing
For Use With (Equipment) 3M™ Secure Click™ HF-800 Series Half Facepiece Reusable Respirators
Includes Head Harness Assembly, Adapter with Port
Material Nylon
Quantity 1 Ea.