Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Secure Click™ Respirator Multi-gas/Vapor Cartridge

Catalog No. 17100817 Shop All 3M Products
Change view
Click to view available options
Quantity:
60 Cartridges/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-100-817 60 Cartridges/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-100-817 Supplier 3M™ Supplier No. 7100153228/DEL

missing translation for 'validMainframeMsg'

The Secure Click™ Multi-Gas/Vapor Cartridge is NIOSH-approved for use against certain organic vapors, sulfur dioxide, hydrogen chloride, chlorine/ammonia/methylamine/formaldehyde, chlorine dioxide, hydrogen fluoride or hydrogen sulfide.

  • Approved for use against organic vapors when used with approved 3M™ Secure Click™ facepieces
  • Secure Click™ cartridges utilize dual flow technology for easy breathing
  • Application: Electro-plating
TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Multi-Gas/Vapor
Certifications/Compliance NIOSH Approved
Connection Type Secure Click
Color Black
Product Type Cartridge
With Exhalation Valve No
For Use With (Equipment) Secure Click Half Facepiece HF-800 Reusable Respirator
Model D8000
Quantity 60 Cartridges/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.