Learn More
Grace Bio Labs Inc SecureSeal™ Imaging Spacers, 100/pk
Catalog No. NC0989329
Encompass_Preferred
SS1X20, 20mm Diameter ID X 0.12mm Depth, 25mm X 25mm OD – 100/pk
Catalog No. NC0989329 Supplier Grace Bio Labs Inc Supplier No. 654006
SS1X20, 20mm Diameter ID X 0.12mm Depth, 25mm X 25mm OD – 100/pk