Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Secure-Seal™ Imaging Spacer, 1 well 20 mm, 0.12 mm Depth
SDP

Catalog No. 5034422
Encompass_Preferred

Confine specimens without compression with SecureSeal imaging spacers. SecureSeal imaging spacers are ultra thin adhesive spacers which peel and stick to coverglass or microscope slides to confine specimens without the need for compression.
Wells: 1 well
Diameter: 20 mm
Depth: 0.12 mm
Dimensions: 25 mm x 25 mm

Catalog No. 50-344-22 Supplier Electron Microscopy Sciences Supplier No. 7032720S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Confine specimens without compression with SecureSeal imaging spacers. SecureSeal imaging spacers are ultra thin adhesive spacers which peel and stick to coverglass or microscope slides to confine specimens without the need for compression.
Wells: 1 well
Diameter: 20 mm
Depth: 0.12 mm
Dimensions: 25 mm x 25 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.