Learn More
Norell Inc 5mm, 800MHz 7" SECURE SERIES NMR TUBE, 5 Tubes per pack INCLUDES "NORLOC WITH SECURITY BAND" CAPS
SDP
Catalog No. NC2832737
Encompass_Preferred
The Secure Series™ NMR tubes feature several patented design elements that add a superior level of sample containment and isolation, safeguarding the integrity of precious or critical NMR samples and assuring secure retention of the NMR tube in the spinner turbine. A patented Security Band™ that engages and locks into the NorLoc II Security Cap™, securely joining the cap to the NMR tube. A patented proprietary surface treatment with a unique textured surface ensures precise and positive retention in Bruker, Agilent/Varian, and Jeol spinner turbines. These tubes are ideal for near room temperature analyses of routine samples exposed only to slight thermal gradients.
Catalog No. NC2832737 Supplier Norell Inc Supplier No. S5508000501780
The Secure Series™ NMR tubes feature several patented design elements that add a superior level of sample containment and isolation, safeguarding the integrity of precious or critical NMR samples and assuring secure retention of the NMR tube in the spinner turbine. A patented Security Band™ that engages and locks into the NorLoc II Security Cap™, securely joining the cap to the NMR tube. A patented proprietary surface treatment with a unique textured surface ensures precise and positive retention in Bruker, Agilent/Varian, and Jeol spinner turbines. These tubes are ideal for near room temperature analyses of routine samples exposed only to slight thermal gradients.