Learn More
Grainger Copeland SecureStart Module Kit, 200/230 VAC, Single-Phase, 1 ea
Catalog No. 502762369
Encompass_Preferred
The SecureStart module kit is a control component designed for use with single-phase permanent split capacitor compressors. This module provides essential motor protection and starting assistance by monitoring voltage supply to prevent motor stalling and optimize performance. It is engineered to detect power interruptions, faulty run capacitors, and reverse operation while helping to mitigate issues such as light dimming or flicker during compressor startup.
  • Compatible with Comfort Alert and CoreSense Diagnostics technology
  • Designed for single-phase compressors with RLA of 32 A or less
  • Includes start capacitor, start relay, and wiring harness
  • Monitors for low voltage and provides compressor protection
  • Features 3-minute restart delay after power interruption
  • RoHS compliant and CE marked
  • Constructed from durable ABS plastic
Catalog No. 50-276-2369 Supplier Grainger Supplier No. 161G60
The SecureStart module kit is a control component designed for use with single-phase permanent split capacitor compressors. This module provides essential motor protection and starting assistance by monitoring voltage supply to prevent motor stalling and optimize performance. It is engineered to detect power interruptions, faulty run capacitors, and reverse operation while helping to mitigate issues such as light dimming or flicker during compressor startup.
  • Compatible with Comfort Alert and CoreSense Diagnostics technology
  • Designed for single-phase compressors with RLA of 32 A or less
  • Includes start capacitor, start relay, and wiring harness
  • Monitors for low voltage and provides compressor protection
  • Features 3-minute restart delay after power interruption
  • RoHS compliant and CE marked
  • Constructed from durable ABS plastic