Learn More
MSC SecureClick Cartridge, 3M, Acid Gas Organic Vapor, D8003, 2 Pack
Catalog No. 5000796124
Encompass
This SecureClick Acid Gas & Organic Vapor cartridge is designed for respiratory protection against organic vapors and acid gases. It features a yellow color code and is part of the D8003 series.
  • Cartridge type for respiratory protection
  • Protects against organic vapor and acid gas
  • Features no NIOSH filter rating
  • Identified by a yellow color code
  • Part of the D8003 manufacturer's series
  • Manufactured in Canada
Catalog No. 50-007-96124 Supplier MSC Supplier No. 18978890
This SecureClick Acid Gas & Organic Vapor cartridge is designed for respiratory protection against organic vapors and acid gases. It features a yellow color code and is part of the D8003 series.
  • Cartridge type for respiratory protection
  • Protects against organic vapor and acid gas
  • Features no NIOSH filter rating
  • Identified by a yellow color code
  • Part of the D8003 manufacturer's series
  • Manufactured in Canada