Learn More
3M™ SecureFit™ 200 Series Anti-Fog Lens
Catalog No. 19070940 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Blue
Clear
Quantity:
Case of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Quantity
19-070-940 Clear Case of 20
19-070-941 Blue Case of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-070-940 Supplier 3M™ Supplier No. 7100180044
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ SecureFit™ 200 Series Anti-Fog Lens provides users with blue temples and lenses to help make eyewear more visible in food preparation and processing areas.
  • Blue temples can be more visible if found in food preparation and processing areas
  • Features proprietary 3M™ Pressure Diffusion Temple Technology which helps diffuse pressure over the ear for a secure, comfortable fitBlue temples can be more visible if found in food preparation and processing areas
  • Applied on both sides of the lens, 3M™ Scotchgard™ Anti-fog is a long-lasting coating that provides better fog and scratch resistance when compared to traditional anti-fog coatings
  • Molded nose bridge temples can be more visible if found in food preparation and processing areas
  • Blue lenses are helpful to reduce glare, eye stress and fatigue in yellow light environments, such as produced by sodium vapor bulbs
  • 3M Catalog# SF201SGAF-BLU , 3M Catalog# SF204SGAF-BLU
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1
Lens Tint Clear
Product Type Protective Eyewear
For Use With (Application) Food and Beverage Manufacturing
Frame Color Blue
Model SF201SGAF-BLU
Quantity Case of 20
No. per Pack 20/Pk.
Temple Style Pressure Diffusion Temple Technology
Lens Material Polycarbonate
Frame Material Polycarbonate
Frame Type Frameless