Learn More
3M™ SecureFit™ 400 Series Protective Eyewear
Catalog No. 19153669 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-153-669 Gray
19-153-666 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-153-669 Supplier 3M™ Supplier No. 7100112244
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ SecureFit™ 400 Series Protective Eyewear with removable foam-lined gasket helps keep debris out of your eyes while providing comfort and cushioning.
  • Stylish design weighs less than 1 oz.
  • Soft adjustable nosepiece to help reduce slippage
  • Incorporates dual-material temples to provide cushioned comfort plus enhanced style and features the proprietary 3M™ Pressure Diffusion Temple Technology – a scientific advancement that helps diffuse pressure over the ear
  • Enhanced temple comfort while not compromising the security of fit across diverse workforces
  • Clear anti-fog coated polycarbonate lens absorbs 99.9% UVA & UVB
  • Removable foam-lined gasket helps keep debris out of your eyes while providing comfort and cushioning.
  • Fits 3M™ SecureFit™ protective eyewear 400-series
  • 3M Catalog# SF401AF-FM , 3M Catalog# SF402AF-FM
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Gray
Product Type Safety Glasses
Frame Color Clear
Model SF402AF-FM
No. per Pack 20/Pk.
Temple Color Black
Lens Material Polycarbonate
Frame Material Plastic
Frame Type Frameless