Learn More
3M™ SecureFit™ 400 Series Safety Glasses
Catalog No. 19070944 Shop All 3M Products
Change view
Click to view available options
Frame Color:
Black, Gray
Black, Green
Black, Yellow
Blue and Gray
Hi-Vis Green and Clear
Light Blue, White
Pink, Black
Yellow, Black
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor
Indoor/Outdoor Mirror
Orange
Lens Coating:
Anti-fog
Scratch-resistant
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19-070-944 Yellow, Black Amber Scratch-resistant
19-070-947 Blue and Gray Indoor/Outdoor Scratch-resistant
19-070-948 Blue and Gray Clear Anti-fog
19-070-949 Blue and Gray Clear Scratch-resistant
19-070-951 Black, Green Gray Anti-fog
19-070-952 Black, Green Clear Anti-fog
17-623-766 Black, Yellow Indoor/Outdoor Mirror Scratch-resistant
17-623-767 Black, Yellow Clear Anti-fog
17-623-768 Black, Yellow Gray Anti-fog
19-070-942 Black, Green Gray Scratch-resistant
19-070-943 Pink, Black Gray -
19-070-945 Black, Gray Orange Scratch-resistant
19-070-946 Light Blue, White Clear -
19-070-950 Hi-Vis Green and Clear Clear -
19-070-953 Black, Green Clear -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 19-070-944 Supplier 3M™ Supplier No. 7100179972
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ SecureFit™ 400 Series Safety Glasses offer models with stylized colors, proprietary Pressure Diffusion Temple (PDT) technology for comfort, and a contoured brow guard to help provide additional coverage from flying particles.
  • Contoured brow guard rests comfortably against forehead and helps provide additional coverage from flying particles
  • Features proprietary 3M™ Pressure Diffusion Temple Technology which helps diffuse pressure over the ear for a secure, comfortable fit
  • Enhanced temple comfort without compromising the security of fit across diverse workforces
  • Soft nosepiece to help reduce slippage
  • Applied on both sides of the lens, 3M™ Scotchgard™ Anti-fog is a long-lasting coating that provides better fog and scratch resistance when compared to traditional anti-fog coatings
  • 3M Catalog# SF401XAF-GRN, SF401XAS-GRN, SF401XSGAF-BLU, SF401XSGAF-GRN, SF401XSGAF-WHT, SF402XAF-GRN, SF402XAS-GRN, SF402XSGAF-BLU, SF403XSGAF-YEL, SF407XSGAF-BLU, SF409XAS-PNK, SF416XAS-BLK
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Amber
Product Type Protective Eyewear
Frame Color Yellow, Black
Model SF403XSGAF-YEL
No. per Pack 20/Pk.
Temple Style Pressure Diffusion Temple Technology
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection Yes
Frame Type Frameless