Learn More
3M™ SecureFit™ 600 Series Protective Eyewear with +1.5 Diopter
Catalog No. 17000697 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
17-000-697 Clear
17-000-700 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-000-697 Supplier 3M™ Supplier No. 7100132357
  • $658.62
    $413.65 / Case of 20
    Save $244.97
    37% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ SecureFit™ 600 Series Protective Eyewear with +1.5 Diopter features proprietary 3M™ Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort while not compromising the security of fit across a diverse workforce.
  • Features proprietary 3M™ Pressure Diffusion Temple Technology which helps diffuse pressure over the ear for a secure, comfortable fit
  • Scotchgard™ Anti-fog Coating lasts longer than traditional anti-fog coatings, helping to increase eyewear performance in wet and steamy environments
  • The proprietary coating retains its effectiveness for at least 25 washings, allowing workers to wear their eyewear longer
  • Workers can feel confident wearing Scotchgard™ Anti-Fog coated eyewear because it meets the requirements of ANSI Z87.1.2015
  • +1.5 diopter combining eye protection with reading enhancement
  • Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated spectacle
  • Grey polycarbonate lens absorbs 99.9% UVA & UVB rays
  • 3M Catalog# SF615GSGAF , 3M Catalog# SF615SGAF
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, UV-treated
Lens Type Magnifying
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Product Type Safety Glasses
Frame Color Gray
Magnification Power +1.50
Model SF615SGAF
No. per Pack 20/Pk.
Temple Style SecureFit™
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Half Frame