Learn More
3M™ SecureFit™ 600 Series Protective Eyewear
Catalog No. 17000692 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor
Low IR
Photochromic
Shade 3
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
17-000-692 Amber
17-000-693 Indoor/Outdoor
17-000-690 Clear
17-000-691 Gray
17-000-694 Low IR
17-000-695 Shade 3
17-000-704 Photochromic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-000-692 Supplier 3M™ Supplier No. 7100141599
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
3M™ SecureFit™ 600 Series Protective Eyewear features proprietary 3M™ Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort while not compromising the security of fit across a diverse workforce.
  • Scotchgard™ Anti-fog Coating lasts longer than traditional anti-fog coatings, helping to increase eyewear performance in wet and steamy environments
  • The proprietary coating retains its effectiveness for at least 25 washings, allowing workers to wear their eyewear longer
  • Workers can feel confident wearing Scotchgard™ Anti-Fog coated eyewear because it meets the requirements of ANSI Z87.1-2015
  • Rugged Anti-Scratch (RAS) coating helps extend the life of the lens
  • Indoor/Outdoor Grey lens helps reduce brightness and glare when working both indoors and outdoors
  • Grey lens are popular in outdoor daytime work environments
  • Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated spectacle
  • Lens absorbs 99.9% UVA & UVB rays
  • 3M Catalog# SF601RAS, SF602RAS, SF603SGAF, SF607SGAF, SF613AS, SF617AS, SF630AS
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, UV-treated
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Amber
Product Type Safety Glasses
Frame Color Gray
Model SF603SGAF
No. per Pack 20/Pk.
Temple Style SecureFit™
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Half Frame