Learn More
3M™ SecureFit™ 600 Series Protective Eyewear
Catalog No. 17000682 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Includes:
—
Foam gasket
Model:
SF601SGAF
SF601SGAF-FM
SF602SGAF
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Includes Model
17-000-682 Clear Foam gasket SF601SGAF-FM
17-000-680 Clear — SF601SGAF
17-000-681 Gray — SF602SGAF
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 17-000-682 Supplier 3M™ Supplier No. 7100133322
  • $513.00
    $319.65 / Case of 20
    Save $193.35
    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal eye protection for diverse workforces. The 3M™ SecureFit™ 600 Series Protective Eyewear features 3M Pressure Diffusion Temple Technology, which provides for a more secure and comfortable fit.
  • Features proprietary 3M Pressure Diffusion Temple Technology which helps diffuse pressure over the ear for a secure, comfortable fit
  • Scotchgard Anti-fog Coating lasts longer than traditional anti-fog coatings, helping to increase eyewear performance in wet and steamy environments
  • The proprietary coating retains its effectiveness for at least 25 washings, allowing workers to wear their eyewear longer
  • Workers can feel confident wearing Scotchgard Anti-Fog coated eyewear because it meets the requirements of ANSI Z87.1.2015
  • Clear polycarbonate lens absorbs 99.9% UVA & UVB
  • 3M Catalog# SF601SGAF , 3M Catalog# SF601SGAF-FM , 3M Catalog# SF602SGAF
Compliance
Meets ANSI Z87.1-2015 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Product Type Protective Eyewear
For Use With (Application) Automotive, Construction, General Manufacturing, Metal Fabrication, Mining, Oil & Gas, Pharmaceutical, Primary Metals
Frame Color Gray
Includes Foam gasket
Model SF601SGAF-FM
Temple Style SecureFit™
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Plastic
UV Protection 99.9% UVA and UVB
Frame Type Half Frame