Learn More
MSC SECUREFIT OTG SAFETY GLS
Catalog No. 5000676192
Encompass
These safety glasses are designed for over-the-glasses wear, offering protection with anti-fog, clear polycarbonate lenses and a durable frameless design.
  • Anti-fog coating improves visibility under hot and humid conditions.
  • Features clear polycarbonate lenses for impact resistance.
  • Frameless design with a blue polycarbonate frame.
  • Over-the-glasses (OTG) style for comfortable wear over prescription eyewear.
  • Equipped with adjustable temples and 3M Pressure Diffusion Temple Technology for a secure fit.
  • Provides UV protection.
  • Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
  • Universal size for broad applicability.
Catalog No. 50-006-76192 Supplier MSC Supplier No. 94428968
These safety glasses are designed for over-the-glasses wear, offering protection with anti-fog, clear polycarbonate lenses and a durable frameless design.
  • Anti-fog coating improves visibility under hot and humid conditions.
  • Features clear polycarbonate lenses for impact resistance.
  • Frameless design with a blue polycarbonate frame.
  • Over-the-glasses (OTG) style for comfortable wear over prescription eyewear.
  • Equipped with adjustable temples and 3M Pressure Diffusion Temple Technology for a secure fit.
  • Provides UV protection.
  • Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
  • Universal size for broad applicability.