Learn More
MSC SECUREFIT SAF GLS/FM GSKT
Catalog No. 5000796720
Encompass
SecureFit 500 Series protective eyewear features a contemporary lens design, adjustable ratchet temples, and Pressure Diffusion Temple (PDT) technology for a personalized fit. This model includes a foam gasket and a gray Scotchgard Protector Anti-Fog and Anti-Scratch lens.
  • Sporty design with contemporary lens shape
  • Adjustable ratchet temples for personalized fit and improved positioning with half-facepiece respirators
  • Flat and flexible temple design for compatibility with protective earmuffs
  • Pressure Diffusion Temple technology
  • U6-rated polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays between 200 nm and 380 nm
  • Scotchgard Protector Anti-Fog coating and Anti-Scratch lens
  • Optional removable foam-lined gasket included
  • Meets the Impact-Rated Protector requirement of the ANSI/ISEA Z87.1-2020 Standard
Catalog No. 50-007-96720 Supplier MSC Supplier No. 17734690
SecureFit 500 Series protective eyewear features a contemporary lens design, adjustable ratchet temples, and Pressure Diffusion Temple (PDT) technology for a personalized fit. This model includes a foam gasket and a gray Scotchgard Protector Anti-Fog and Anti-Scratch lens.
  • Sporty design with contemporary lens shape
  • Adjustable ratchet temples for personalized fit and improved positioning with half-facepiece respirators
  • Flat and flexible temple design for compatibility with protective earmuffs
  • Pressure Diffusion Temple technology
  • U6-rated polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays between 200 nm and 380 nm
  • Scotchgard Protector Anti-Fog coating and Anti-Scratch lens
  • Optional removable foam-lined gasket included
  • Meets the Impact-Rated Protector requirement of the ANSI/ISEA Z87.1-2020 Standard