Learn More
3M™ SecureFit™ Safety Glasses
Catalog No. 19040636 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Gray
Catalog No. Lens Tint
19-040-636 Gray
1 options
This item is not returnable. View return policy
Catalog No. 19-040-636 Supplier 3M™ Supplier No. 7010342616
Add to Cart
Add to Cart
This item is not returnable. View return policy
SecureFit™ Safety Glasses feature proprietary 3M™ Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort while not compromising the security of fit across a diverse workforce.
  • 3M Pressure Diffusion Technology allows temples to naturally adjust to individual head sizes eliminating the need to stock multiple frame sizes at worksites
  • Secure, snug fit keeps eyewear in place without slippage even when the person is in motion
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380 nm and meet the requirements of CSA Z94.3 and ANSI Z87.1-2015
  • SecureFit™ Alternate fit protective eyewear expands the fit of eyewear to multiple face shapes and sizes with unique design adjustments such as lens angling angled to help accommodate high cheekbone structure and soft nose pads designed to help fit securely on a low nose bridge.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3
Lens Tint Gray
Product Type Safety Glasses
For Use With (Application) Assembly, Masonary, UV Protection, Sawing, Sanding, Riveting, Remediation, Power Fastening, Pouring/Casting, Painting, Machining, Chipping, Machine Operations, Grinding, Facility Maintenance, Electrical, Drilling, Demolition, Cleaning, Chiseling, Welding
Frame Color Gray
No. per Case 20/Cs.
Model SF302AF
Temple Style 3M Pressure Diffusion Temple Technology
Temple Color Gray
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Frameless