Learn More
MSC SECUREFIT SAFETY GLS
Catalog No. 5000793821
Encompass
These safety glasses feature amber polycarbonate lenses with anti-fog and scratch-resistant coatings, designed for optimal visibility in various conditions. The frameless design incorporates 3M Pressure Diffusion Temple Technology for enhanced comfort and meets ANSI Z87.1-2015 standards.
  • Tinted glasses style
  • Lens coating: anti-fog and scratch-resistant
  • Frameless, wrap around design
  • Amber lens color
  • Polycarbonate lens material
  • UV protection: yes
  • Meets ANSI Z87.1-2015 standards
Catalog No. 50-007-93821 Supplier MSC Supplier No. 46722211
These safety glasses feature amber polycarbonate lenses with anti-fog and scratch-resistant coatings, designed for optimal visibility in various conditions. The frameless design incorporates 3M Pressure Diffusion Temple Technology for enhanced comfort and meets ANSI Z87.1-2015 standards.
  • Tinted glasses style
  • Lens coating: anti-fog and scratch-resistant
  • Frameless, wrap around design
  • Amber lens color
  • Polycarbonate lens material
  • UV protection: yes
  • Meets ANSI Z87.1-2015 standards