Learn More
MSC SECUREFIT SAFETY GLS
Catalog No. 5000685862
Encompass
3M SecureFit 400 Series Safety Glasses, model SF407SGAF-BLU, feature Indoor/Outdoor Gray Scotchgard Anti-Fog and Anti-Scratch lenses. These frameless safety glasses are designed with 3M Pressure Diffusion Temple Technology for a comfortable and secure fit, meeting ANSI Z87.1-2015 and CSA Z94.3 standards.
  • Pressure Diffusion Temple Technology provides comfortable pressure equalization.
  • Curved temples apply pressure to the less sensitive back of the head.
  • U6 rated polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays.
  • Anti-fog coating improves visibility in hot and humid conditions.
  • Soft, adjustable nose pads reduce slippage and personalize fit.
  • Wraparound design offers a wide field of vision.
Catalog No. 50-006-85862 Supplier MSC Supplier No. 46722245
3M SecureFit 400 Series Safety Glasses, model SF407SGAF-BLU, feature Indoor/Outdoor Gray Scotchgard Anti-Fog and Anti-Scratch lenses. These frameless safety glasses are designed with 3M Pressure Diffusion Temple Technology for a comfortable and secure fit, meeting ANSI Z87.1-2015 and CSA Z94.3 standards.
  • Pressure Diffusion Temple Technology provides comfortable pressure equalization.
  • Curved temples apply pressure to the less sensitive back of the head.
  • U6 rated polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays.
  • Anti-fog coating improves visibility in hot and humid conditions.
  • Soft, adjustable nose pads reduce slippage and personalize fit.
  • Wraparound design offers a wide field of vision.