Learn More
MSC SECUREFIT SAFETY GLS
Catalog No. 5000796521
Encompass
The 3M SecureFit 500 Series Safety Glasses provide advanced eye protection with a contemporary design. Integrating 3M Pressure Diffusion Temple (PDT) technology and adjustable ratchet temples, they ensure a personalized, comfortable fit. This SF502SGAF-BLK model features a grey lens with Scotchgard Anti-Fog and anti-scratch coatings for clear vision. U6-rated polycarbonate lenses block 99.9% of UVA, UVB, and UVC rays.
  • Pressure Diffusion Temple (PDT) technology provides comfortable, self-adjusting fit
  • Adjustable ratchet temples for customizable lens angle and position
  • Grey lens with Scotchgard Anti-Fog and anti-scratch coating
  • U6-rated polycarbonate lenses block 99.9% of UVA, UVB, and UVC rays
  • Lightweight, low-profile frame with curved temple arms
  • Compatible with protective earmuffs
  • Meets ANSI Z87.1-2020 and AS/NZS 1337.1:2010 standards
Catalog No. 50-007-96521 Supplier MSC Supplier No. 17734401
The 3M SecureFit 500 Series Safety Glasses provide advanced eye protection with a contemporary design. Integrating 3M Pressure Diffusion Temple (PDT) technology and adjustable ratchet temples, they ensure a personalized, comfortable fit. This SF502SGAF-BLK model features a grey lens with Scotchgard Anti-Fog and anti-scratch coatings for clear vision. U6-rated polycarbonate lenses block 99.9% of UVA, UVB, and UVC rays.
  • Pressure Diffusion Temple (PDT) technology provides comfortable, self-adjusting fit
  • Adjustable ratchet temples for customizable lens angle and position
  • Grey lens with Scotchgard Anti-Fog and anti-scratch coating
  • U6-rated polycarbonate lenses block 99.9% of UVA, UVB, and UVC rays
  • Lightweight, low-profile frame with curved temple arms
  • Compatible with protective earmuffs
  • Meets ANSI Z87.1-2020 and AS/NZS 1337.1:2010 standards