Learn More
MSC SECUREFIT SAFETY GLS
Catalog No. 5000796454
Encompass
These safety glasses from the SecureFit 500 Series feature anti-fog and scratch-resistant polycarbonate lenses, designed to improve visibility in hot and humid conditions. They offer a frameless, wrap-around design with adjustable temples for a comfortable and secure fit.
  • Tinted lenses
  • Anti-fog and scratch-resistant coating
  • Frameless, wrap-around design
  • Brown lens color
  • Olive frame color
  • Universal size
  • Polycarbonate lens material
  • Adjustable temples
  • UV protection
  • Meets ANSI Z87.1-2010 standards
  • Ratchet temple type
  • Removable gasket
Catalog No. 50-007-96454 Supplier MSC Supplier No. 17734583
These safety glasses from the SecureFit 500 Series feature anti-fog and scratch-resistant polycarbonate lenses, designed to improve visibility in hot and humid conditions. They offer a frameless, wrap-around design with adjustable temples for a comfortable and secure fit.
  • Tinted lenses
  • Anti-fog and scratch-resistant coating
  • Frameless, wrap-around design
  • Brown lens color
  • Olive frame color
  • Universal size
  • Polycarbonate lens material
  • Adjustable temples
  • UV protection
  • Meets ANSI Z87.1-2010 standards
  • Ratchet temple type
  • Removable gasket