Learn More
MSC SECUREFIT SAFETY GLS
Catalog No. 5000789721
Encompass
The 3M SecureFit 100 Series safety glasses offer comfortable, economical eye protection with an innovative temple design that self-adjusts to the wearer's head size. Curved temples apply pressure to the less sensitive back of the head, ensuring a secure fit even during rapid head movement.
  • Innovative temple design with slotted ribs for pressure equalization
  • Temples flex and self-adjust to head size
  • Sporty design with contemporary lens shape
  • Flat and flexible temple design for compatibility with protective earmuffs
  • Polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays
  • Meets ANSI-ISEA Z87.1-2020 Impact-Rated Protector requirement
  • Features Pressure Diffusion Temple Technology
  • Anti-scratch and anti-fog coated gray lenses
  • Optical class 1 lens for prolonged use
  • Weighs approximately 21 g (0.7 oz)
Catalog No. 50-007-89721 Supplier MSC Supplier No. 17734633
The 3M SecureFit 100 Series safety glasses offer comfortable, economical eye protection with an innovative temple design that self-adjusts to the wearer's head size. Curved temples apply pressure to the less sensitive back of the head, ensuring a secure fit even during rapid head movement.
  • Innovative temple design with slotted ribs for pressure equalization
  • Temples flex and self-adjust to head size
  • Sporty design with contemporary lens shape
  • Flat and flexible temple design for compatibility with protective earmuffs
  • Polycarbonate lenses absorb 99.9% of UVA, UVB, and UVC rays
  • Meets ANSI-ISEA Z87.1-2020 Impact-Rated Protector requirement
  • Features Pressure Diffusion Temple Technology
  • Anti-scratch and anti-fog coated gray lenses
  • Optical class 1 lens for prolonged use
  • Weighs approximately 21 g (0.7 oz)