Learn More
MSC SECUREFIT SAFETY GLS
Catalog No. 5000789722
Encompass
3M SecureFit 100 Series Safety Glasses feature a sleek contemporary lens design with proprietary Pressure Diffusion Temple Technology, providing enhanced comfort and security at an economical price point.
  • Pressure Diffusion Temple (PDT) technology for a secure fit
  • Curved temples for enhanced security
  • Anti-scratch or anti-fog/anti-scratch coatings available
  • Absorbs 99.9% UVA, UVB, and UVC rays (200-380 nm)
  • Sleek, sporty lens shape
  • Molded nose bridge for comfort
  • Lightweight design, less than 1 oz.
Catalog No. 50-007-89722 Supplier MSC Supplier No. 17734419
3M SecureFit 100 Series Safety Glasses feature a sleek contemporary lens design with proprietary Pressure Diffusion Temple Technology, providing enhanced comfort and security at an economical price point.
  • Pressure Diffusion Temple (PDT) technology for a secure fit
  • Curved temples for enhanced security
  • Anti-scratch or anti-fog/anti-scratch coatings available
  • Absorbs 99.9% UVA, UVB, and UVC rays (200-380 nm)
  • Sleek, sporty lens shape
  • Molded nose bridge for comfort
  • Lightweight design, less than 1 oz.