Learn More
MSC SECUREFIT SAFETY GLS
Catalog No. 5000789732
Encompass
These safety glasses feature anti-fog and scratch-resistant gray polycarbonate lenses with a frameless, wrap-around design. They offer UV protection and meet ANSI Z87.1-2010 standards.
  • Anti-fog coatings improve visibility
  • Scratch-resistant polycarbonate lenses
  • Frameless, wrap-around design
  • Adjustable temples
  • UV protection
  • Meets ANSI Z87.1-2010 standards
  • Pressure Diffusion Temple Technology
Catalog No. 50-007-89732 Supplier MSC Supplier No. 17734625
These safety glasses feature anti-fog and scratch-resistant gray polycarbonate lenses with a frameless, wrap-around design. They offer UV protection and meet ANSI Z87.1-2010 standards.
  • Anti-fog coatings improve visibility
  • Scratch-resistant polycarbonate lenses
  • Frameless, wrap-around design
  • Adjustable temples
  • UV protection
  • Meets ANSI Z87.1-2010 standards
  • Pressure Diffusion Temple Technology