Learn More
Research Products International Corp SecureSeal Imaging Spacer [Chambers], 13mm Dia, 0.12mm Thick, 100/PK
SDP
Catalog No. 501977603
Encompass
Imaging spacers are ultra-thin adhesive spacers which peel-and-stick to coverglass or microscope slides to confine specimens without compression. Layer multiple spacers to custom build chambers to any depth desired.
Catalog No. 50-197-7603 Supplier Research Products International Corp Supplier No. 247462
Imaging spacers are ultra-thin adhesive spacers which peel-and-stick to coverglass or microscope slides to confine specimens without compression. Layer multiple spacers to custom build chambers to any depth desired.