Learn More
Electron Microscopy Sciences Held Secure Slide In/Out Cards Orange
SDP
Catalog No. 501928285
Encompass_Preferred
A durable corrugated filing box kit for the proper filing, identification, and storage of glass slides or cassettes. It holds up to 2,000 slides or 500+ cassettes per box and has partitioned drawers for easy retrieval. Features
Double tabbed in back for additional safety.
Printing on box front designed for easy inventory identification.
Internal corrugated construction designed for multiple stacking.
50 In/Out Cards come with each Slide/Cassette case for slide retrieval documentation.
25 Index Cards come with each Slide Kit for slide retrieval documentation.
Internal corrugated construction supports up to 250 lbs and the tray remains easily accessible.
The complete set comes unassembled in packages of 10.
Manufactured in the USA.
Catalog No. 50-192-8285 Supplier Electron Microscopy Sciences Supplier No. 6328052OR
A durable corrugated filing box kit for the proper filing, identification, and storage of glass slides or cassettes. It holds up to 2,000 slides or 500+ cassettes per box and has partitioned drawers for easy retrieval. Features
Double tabbed in back for additional safety.
Printing on box front designed for easy inventory identification.
Internal corrugated construction designed for multiple stacking.
50 In/Out Cards come with each Slide/Cassette case for slide retrieval documentation.
25 Index Cards come with each Slide Kit for slide retrieval documentation.
Internal corrugated construction supports up to 250 lbs and the tray remains easily accessible.
The complete set comes unassembled in packages of 10.
Manufactured in the USA.