Learn More
Research Products International Corp SecureSlip Glass Coverslips, 12 x 12 mm, Sterile, 75 per Package
SDP
Catalog No. 50212490
Encompass
Individual 12 mm square glass coverslips with a silicone backing. Thin non-cytotoxic silicone layer bonds coverslip to any smooth surface on contact. Transparent silicone will not interfere with cells during culture. Up to fifteen coverslips can be placed in a culture plate. Remove easily from plate by peeling away using forceps. Each sterile pouch contains fifteen silicone backed coverslips, seventy-five per package. Easy to follow instruction sheet included.
Catalog No. 50-212-490 Supplier Research Products International Corp Supplier No. 244224
Individual 12 mm square glass coverslips with a silicone backing. Thin non-cytotoxic silicone layer bonds coverslip to any smooth surface on contact. Transparent silicone will not interfere with cells during culture. Up to fifteen coverslips can be placed in a culture plate. Remove easily from plate by peeling away using forceps. Each sterile pouch contains fifteen silicone backed coverslips, seventy-five per package. Easy to follow instruction sheet included.