Learn More
Grainger Security Screwdriver Set Torx(R) 7 pcs.
Catalog No. 502763690
Encompass_Preferred
The Grainger 170G07 is a Westward brand 7-inch high carbon steel straight snips that can cut up to 18 gauge mild steel, 20 gauge stainless steel, or 16 gauge aluminum. It has precision ground blades for clean cuts and soft grip handles for comfort. This tool weighs 0.75 lbs and is 12 inches long. It can be used for straight cutting or trimming sheet metal.
Catalog No. 50-276-3690 Supplier Grainger Supplier No. 170G07
The Grainger 170G07 is a Westward brand 7-inch high carbon steel straight snips that can cut up to 18 gauge mild steel, 20 gauge stainless steel, or 16 gauge aluminum. It has precision ground blades for clean cuts and soft grip handles for comfort. This tool weighs 0.75 lbs and is 12 inches long. It can be used for straight cutting or trimming sheet metal.