Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 150mm Length with 10-32 Fittings

Catalog No. 502400416 Shop All Phenomenex Products
Encompass_Preferred

SecurityLINK PEEK-Lined SS 100m ID AJ1-3421

Catalog No. 50-240-0416 Supplier Phenomenex Inc Supplier No. AJ13421
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SecurityLINK PEEK-Lined SS 100m ID AJ1-3421

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.