Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 350mm Length with 10-32 Fittings

Catalog No. 502398628 Shop All Phenomenex Products
Encompass_Preferred

SecurityLINK PEEK-Lined SS 75m ID AJ1-3361

Catalog No. 50-239-8628 Supplier Phenomenex Inc Supplier No. AJ13361
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SecurityLINK PEEK-Lined SS 75m ID AJ1-3361

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.