Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 150mm Length with Single 10-32 Fitting

Catalog No. 502398984 Shop All Phenomenex Products
Encompass_Preferred

SecurityLINK PEEKsil 25m ID AJ1-2124

Catalog No. 50-239-8984 Supplier Phenomenex Inc Supplier No. AJ12124
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SecurityLINK PEEKsil 25m ID AJ1-2124

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.