Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc SecurityLINK PEEKsil 50µm ID

Catalog No. 502397973 Shop All Phenomenex Products
Encompass_Preferred

SecurityLINK PEEKsil 50µm ID 200mm Length with 10-32 Fittings

Catalog No. 50-239-7973 Supplier Phenomenex Inc Supplier No. AJ12231
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SecurityLINK PEEKsil 50µm ID 200mm Length with 10-32 Fittings

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.