Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc SecurityLINK SS 100µm ID

Catalog No. 502397576 Shop All Phenomenex Products
Encompass_Preferred

SecurityLINK SS 100µm ID 100mm Length with 10-32 Fittings

Catalog No. 50-239-7576 Supplier Phenomenex Inc Supplier No. AJ11411
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SecurityLINK SS 100µm ID 100mm Length with 10-32 Fittings

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.