Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 350mm Length with 10-32 Fittings

Catalog No. 502396852 Shop All Phenomenex Products
Encompass_Preferred

SecurityLINK SS 125m ID AJ1-1561

Catalog No. 50-239-6852 Supplier Phenomenex Inc Supplier No. AJ11561
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SecurityLINK SS 125m ID AJ1-1561

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.