Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc SecurityLINK SS 254µm ID

Catalog No. 502398063 Shop All Phenomenex Products
Encompass_Preferred

SecurityLINK SS 254µm ID 600mm Length with 10-32 Fittings

Catalog No. 50-239-8063 Supplier Phenomenex Inc Supplier No. AJ11681
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SecurityLINK SS 254µm ID 600mm Length with 10-32 Fittings

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.