Learn More
PDC Securline™ Blood Barcode Narrow Wristband with Barcode Labels
Description
- Wristband labels with large alphanumeric font (14 pt.) and barcodes
- Total of 20 matching labels (8 alphanumeric on wristband; 12 barcode/alphanumeric on draw tube tail)
- Narrow style wristband to fit adults or pediatric patients
- Streamlines the blood transfusion process and improves patient safety by reducing human errors related to patient identification
- Saves time and money by streamlining the patient ID and blood transfusion workflow and minimizes paperwork
- Integrates seamlessly with your existing blood bank system
- Patented dual-purpose, hybrid style lets you handwrite information or use with your LIS label
- Standardize and consolidate to a single blood band, even if you have multiple labs
- Designed for inpatient or outpatient use
- Laminate shield protects patient information and bar code from water or solvents for accurate bar code scanning
- Advanced material resists moisture, alcohol and hand sanitizer to preserve bar code integrity for accurate scanning
- Meets The Joint Commission's National Patient Safety Goal for blood transfusions for a one-person verification system
Specifications
Specifications
| Includes | 100/Box |
| Closure Type | Snap |
| Material | Polypropylene, PET |
| Color | Red |
| Product Type | Blood Band |
| Dimensions (L x W) | 0.6 x 11 in. |