Learn More
PDC Securline™ Blood Barcode Wristband, Tail with Barcode and Alpha Numeric Labels
Catalog No. 22138371
Change view
Click to view available options
Color:
Green
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-138-371 Red
22-138-353 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-138-371 Supplier PDC Supplier No. 243316PDB
Add to Cart
Add to Cart
Using bar code and serialized alphanumeric codes, the Securline™ Bar Code Blood Band accurately matches the right patient to the right blood, ensuring maximum safety throughout the blood transfusion process.
  • Wristband labels with large alphanumeric font (14 pt.) and barcodes
  • Total of 20 matching labels (8 alphanumeric and barcode on wristband; 12 barcode/alphanumeric on draw tube tail)
  • Streamlines the blood transfusion process and improves patient safety by reducing human errors related to patient identification
  • Saves time and money by streamlining the patient ID and blood transfusion workflow and minimizes paperwork
  • Integrates seamlessly with your existing blood bank system
  • Patented dual-purpose, hybrid style lets you handwrite information or use with your LIS label
  • Standardize and consolidate to a single blood band, even if you have multiple labs
  • Designed for inpatient or outpatient use
  • Laminate shield protects patient information and bar code from water or solvents for accurate bar code scanning
  • Advanced material resists moisture, alcohol and hand sanitizer to preserve bar code integrity for accurate scanning
  • Meets The Joint Commission's National Patient Safety Goal for blood transfusions for a one-person verification system
Specifications
Includes 100/Box
Closure Type Snap
Material Polypropylene, PET
Color Red
Product Type Blood Band
Dimensions (L x W) 3.25 x 11 in.