Learn More
PDC Securline™ VeriDigit™ Bar Code Blood Wristband, Light Blue
Catalog No. 21235784
Change view
Click to view available options
Product Type:
Barcode Blood Band
Catalog No. Product Type
21-235-784 Barcode Blood Band
1 options
Catalog No. 21-235-784 Supplier PDC Supplier No. 243718PDB
Add to Cart
Add to Cart
Light blue wristband for safe bedside transfusion, supporting patient ID and easy reattachment with matching bar code labels.
  • Integrates with Cerner Bridge Transfusion Administration system for accurate patient verification
  • Permanent high-contrast bar code ensures reliable identity confirmation
  • Alerts staff if an incorrect bar code is scanned to prevent errors
  • Embedded prefix and suffix enhance workflow validation and safety checks
  • Easy-to-read ID numbers with GS1-128 bar coding for healthcare scanner compatibility
  • Peel-off adhesive tail with matching bar code labels for draw tubes
  • Reattachable with Xtender™ Band for patient convenience
  • Suitable for adult and pediatric patients with clear, visible identification
Specifications
Closure Type Snap
Color Blue
Product Type Barcode Blood Band