Learn More
PDC Securline™ VeriDigit™ Barcode Blood Wristband, Serialized Barcode Write-On/Label
Catalog No. 22138364
Change view
Click to view available options
Color:
Red
Catalog No. Color
22-138-364 Red
1 options
Catalog No. 22-138-364 Supplier PDC Supplier No. 243816PDB
Add to Cart
Add to Cart
The Securline™ VeriDigit™ Blood Band Wristband is specially designed to support the Cerner Bridge Transfusion Administration system. VeriDigit™ helps ensure positive patient identification prior to transfusion at the patient’s bedside.
  • Features a special permanent bar code that must be scanned by the transfusionist as part of the Cerner Transfusion Administration workflow
  • The permanent bar code ID contains an embedded prefix and suffix code when scanned, ensuring staff verified the patient at bedside
  • Large, easy-to-read blood band ID numbers and crisp bar codes
  • GS1-128 bar coding symbology
  • Adhesive peel-off tail with matching bar code labels applies directly to draw tubes
Specifications
Includes 100/Box
Closure Type Snap
Material Polypropylene, PET
Color Red
Product Type Blood Band
Dimensions (L x W) 0.6 x 11 in.