Learn More
PDC Securlink Blood Band
Catalog No. 22138342
Change view
Click to view available options
Includes:
250 Bands
Catalog No. Includes
22-138-342 250 Bands
1 options
Catalog No. 22-138-342 Supplier PDC Supplier No. 213600PDJ
Add to Cart
Add to Cart
Bloodband only - part of Securlink™ Bloodband System
  • The Securlink™ System links the patient, the transfused blood and all the related paperwork to assure safe and complete blood transfusion.
  • The wristband is made of soft vinyl, featuring a tamper proof patented SecurSnap™ closure.
  • Designed for inpatient or outpatient use
  • Plastic material resists moisture, alcohol and hand sanitizer
Specifications
Includes 250 Bands
Closure Type Snap
Material Polypropylene, PET
Color Red
Product Type Blood Band
Dimensions (L x W) 4 x 0.3 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov